
Ammoniumhexachlorstannat(IV) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16960-53-5
:
Formula:H₈Cl₆N₂Sn
MDL Number:MFCD00036340
:367.5 g/mol
Melting Pt: Dec.
Density:2,4
EINECS:241-033-2
:GHS07
Storage Temperature:Stue temperatur
Related Information
Frequently Bought Together