3-Bromobenzoic acid
Catalog # MOLEM34821704
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: m-Bromobenzoic Acid
:
:201.02 g/mol
Boiling Pt:308 °C (1013 hPa)
Boiling Pt:308 °C (1013 hPa)
Melting Pt:155 °C
Density:1,845 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-562-3
:GHS07
CAS Number:585-76-2
Formula:C₇H₅BrO₂
MDL Number:MFCD00002487