Order Entry
Denmark
Orders LinkContactUsLinkComponent
Intravenous catheters, Surflash™
Supplier:   TERUMO
Intravenous catheters, Surflash™
Supplier:  TERUMO
Certificates
Intravenous catheters, Surflash™
8 Options Available Below
Product Details & Documents
Surflash is made of PU, which is firm at room temperature but soft at body temperature, making the catheter more comfortable.
  • Only exists in the straight version
  • Enables quick and easy confirmation of the placement in the vein by an additional flashback
  • Higher certainty of proper placement
Helpful with patients whose veins are hard to detect and in which venipuncture is more difficult, such as small children, people undergoing chemotherapy.
Product Family Options
Product Information
  • Gauge
    Colour
    Material
    Length
    Availability
    Price
  • TERUSR-FF1464
    Intravenous catheters
    Intravenous catheters
    G 14
    Orange
    PU
    64 mm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 811,00 kr
    811,00 kr
    1 * 50 Items
       811,00 kr
  • TERUSR-FF1651
    Intravenous catheters
    Intravenous catheters
    G 16
    Grey
    PU
    51 mm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 716,00 kr
    716,00 kr
    1 * 50 Items
       716,00 kr
  • TERUSR-FF1832
    Intravenous catheters
    Intravenous catheters
    G 18
    Green
    PU
    32 mm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 716,00 kr
    716,00 kr
    1 * 50 Items
       716,00 kr
  • TERUSR-FF1851
    Intravenous catheters
    Intravenous catheters
    G 18
    Green
    PU
    51 mm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 716,00 kr
    716,00 kr
    1 * 50 Items
       716,00 kr
  • TERUSR-FF2032
    Intravenous catheters
    Intravenous catheters
    G 20
    Pink
    PU
    32 mm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 716,00 kr
    716,00 kr
    1 * 50 Items
       716,00 kr
  • TERUSR-FF2051
    Intravenous catheters
    Intravenous catheters
    G 20
    Pink
    PU
    51 mm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 716,00 kr
    716,00 kr
    1 * 50 Items
       716,00 kr
  • TERUSR-FF2225
    Intravenous catheters
    Intravenous catheters
    G 22
    Blue
    PU
    25 mm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 716,00 kr
    716,00 kr
    1 * 50 Items
       716,00 kr
  • TERUSR-FF2419
    Intravenous catheters
    Intravenous catheters
    G 24
    Yellow
    PU
    19 mm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 716,00 kr
    716,00 kr
    1 * 50 Items
       716,00 kr