Intravenous catheter, Surflo®
: TERUMO
This IV catheter exists in three types: Straight (EFTE), wings (FEP), and wings and injection port (FEP).
- Catheter tip is perfectly tapered to ensure a smooth transition from needle to catheter
- Because the material is stronger than other materials, there is less likeliness of split tips and peelback
- For almost each gauge an obturator is available with the same colour as the catheter
- Obturator prevents thrombus formation in the lumen of the catheter when the infusion is discontinued
This IV catheter exists in three types: Straight (EFTE), wings (FEP), and wings and injection port (FEP).
- Catheter tip is perfectly tapered to ensure a smooth transition from needle to catheter
- Because the material is stronger than other materials, there is less likeliness of split tips and peelback
- For almost each gauge an obturator is available with the same colour as the catheter
- Obturator prevents thrombus formation in the lumen of the catheter when the infusion is discontinued
Product Family Options
Straight
- DescriptionGaugeColourMaterialLengthØ×L
- Straight cannulaeG 24YellowETFE0,67×19 mm1 * 50 Items589,00 kr
- Straight cannulaeG 22BlueETFE0,85×25 mm1 * 50 Items589,00 kr
- Straight cannulaeG 20PinkETFE1,10×32 mm1 * 50 Items589,00 kr
- Straight cannulaeG 20PinkETFE1,10×51 mm1 * 50 Items589,00 kr
- Straight cannulaeG 18GreenETFE32 mm1,30×32 mm1 * 50 Items589,00 kr
- Straight cannulaeG 18GreenETFE51 mm1,30×51 mm1 * 50 Items589,00 kr
- Straight cannulaeG 18GreenETFE64 mm1,30×64 mm1 * 50 Items953,00 kr
- Straight cannulaeG 16GreyETFE51 mm1,70×51 mm1 * 50 Items589,00 kr
- Straight cannulaeG 16GreyETFE64 mm1,70×64 mm1 * 50 Items953,00 kr
- Straight cannulaeG 14OrangeETFE64 mm2,15×64 mm1 * 50 Items600,00 kr