Order Entry
Denmark
ContactUsLinkComponent
Microplates, 1536-well
  391-1972
undefined
Microplates, 1536-well
  391-1972
  • Description:
    Microplate, PP, 1536-well, flat bottom with fixed glass inserts, chemically inert
  • Pk:
    1
  • Well volume:
    50 µl
Microtitration plate, made from PP, with fixed glass inserts.
  • High purity and chemically inert
  • 1536 wells
  • Translucent plastic cover included in standard scope of delivery
Plate dimensions: 124×81 mm