Order Entry
Denmark
Orders LinkContactUsLinkComponent
Clamps
Clamps
Supplier:   Lenz Laborglas
Clamps
Supplier:  Lenz Laborglas
Certificates
Clamps
3 Options Available Below
Product Details & Documents
Stainless steel.
  • Excellent chemical resistance
  • Excellent temperature resistance
For fixing lids onto reaction vessels, available for flat flanges LF100-200.
Product Family Options
Product Information
  • For
    Availability
    Price
  • Flat flanges DN 100
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 12.880,00 kr
    12.880,00 kr
    1 * 1 Items
       12.880,00 kr
  • Flat flanges DN 150
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 14.070,00 kr
    14.070,00 kr
    1 * 1 Items
       14.070,00 kr
  • Flat flanges DN 200
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 19.640,00 kr
    19.640,00 kr
    1 * 1 Items
       19.640,00 kr