


2-Methyl-1-penten ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:763-29-1
:
Formula:C₆H₁₂
MDL Number:MFCD00009405
:84.16 g/mol
Boiling Pt:61°C
Melting Pt:–136°C
Density:0,7083 g/cm³ (0 °C)
Flash Pt:–28°C
Storage Temperature:Stue temperatur
EINECS:212-108-7
UN:2288
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together