Oil sorbents
Supplier: 3M
Jump to:
About this item
Oil adsorbent sheets or pads on a roll for spill control.
- Hydrophobic
- Oil (not water) is recovered
- Can be used outside as well as in-plant
4 Options Available Below
Product Details & Documents
Oil adsorbent sheets or pads on a roll for spill control.
- Hydrophobic
- Oil (not water) is recovered
- Can be used outside as well as in-plant
Product Family Options
Product Information
- AbsorbencyL×WAvailabilityPrice
- MMMAHP156Polyester sorbent sheets with charge dissipating PP scrim on one sidePolyester sorbent sheets with charge dissipating PP scrim on one side142 litres/case480×430 mmLog in for availability1 * 100 Items1.410,00 kr
- MMMAHP100Polyester sorbent with charge dissipating PP scrim on one sidePolyester sorbent with charge dissipating PP scrim on one side276 litres/roll44 m × 960 mmLog in for availability1 * 1 ROL3.070,00 kr
- MMMAT150PP sorbent roll
- MMMATF2001Sorbent rolls