Order Entry
Denmark
Orders LinkContactUsLinkComponent
Powder funnels
Powder funnels
Supplier:   HIRSCHMANN
Powder funnels
Supplier:  HIRSCHMANN
Certificates
Powder funnels
5 Options Available Below
Product Details & Documents
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
Product Family Options
Product Information
  • Funnel O-Ø
    Length
    Stem length
    Availability
    Price
  • HIRS7760101
    15 mm stem I-Ø
    15 mm stem I-Ø
    65 mm
    68 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 226,00 kr
    226,00 kr
    1 * 10 Items
       226,00 kr
  • HIRS7760102
    21 mm stem I-Ø
    21 mm stem I-Ø
    80 mm
    76 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 317,00 kr
    317,00 kr
    1 * 10 Items
       317,00 kr
  • HIRS7760104
    26 mm stem I-Ø
    26 mm stem I-Ø
    120 mm
    110 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 408,00 kr
    408,00 kr
    1 * 10 Items
       408,00 kr
  • HIRS7760105
    28 mm stem I-Ø
    28 mm stem I-Ø
    150 mm
    138 mm
    22 mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 275,00 kr
    275,00 kr
    1 * 5 Items
       275,00 kr
  • HIRS7760103
    22 mm stem I-Ø
    22 mm stem I-Ø
    100 mm
    94 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 325,00 kr
    325,00 kr
    1 * 10 Items
       325,00 kr