Order Entry
Denmark
Orders LinkContactUsLinkComponent
3-Methylpentane ≥99%

3-Methylpentane ≥99%

Flammable
Health
Irritant
Environment
3-Methylpentane ≥99%
Certificates
3-Methylpentane ≥99%
3-Methylpentane ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99+% CAS No.: 96-14-0 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -32°(-26°F) Refractive Index: 1.3770 Odour: Petroleum-like Melting Point: -118° Boiling Point: 63-65° Beilstein Reference: 1730734 UN No.: 1208 MDL No.: MFCD00009342

  • CAS Number:96-14-0
  • Formula:C₆H₁₄
  • MDL Number:MFCD00009342
  • Molecular Weight:86.18 g/mol
  • Boiling Pt:63…65 °C
  • Melting Pt:–118 °C
  • Density:0.663
  • Flash Pt:–32 °C (–26 °F)
  • Storage Temperature:Ambient
  • EINECS:202-481-4
  • UN:1208
  • ADR:3,II
Product Family Options

Product Information