Order Entry
Denmark
Orders LinkContactUsLinkComponent
3-Methylpentane 99+%

3-Methylpentane 99+%

Irritant
Health
Environment
Flammable
3-Methylpentane 99+%
Certificates
3-Methylpentane 99+%

3 Options Available Below

Product Details & Documents
  • CAS Number:96-14-0
  • Formula:C₆H₁₄
  • MDL Number:MFCD00009342
  • Molecular Weight:86.18 g/mol
  • Boiling Pt:64 °C (760 mmHg)
  • Melting Pt:–118 °C
  • Density:0.66 g/cm³
  • Flash Pt:–32 °C
  • Storage Temperature:Ambient
  • EINECS:202-481-4
  • UN:1208
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3755 to 1.3775 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Assay:
    ≤8 % (Tetracyclin hydrochloride) On dry substance
  • Water:
    ≤2.0 %
  • Sulfated ash:
    ≤0.5 %
  • Identification:
    Passes test
  • pH:
    2.3 to 3.3 (1 % in water CO2 free)
  • Specific optical rotation:
    -235° to -250° (20°C, 589 nm) (c=0.25, water) (on dry substance)
  • Absorbance:
    ≤0.40 (460 nm)
  • Heavy metals (as Pb):
    ≤50 ppm
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.