2-Methylpentane, Supelco®
68310-50ML
: Merck
:
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
:
:62 °C (1013 hPa)
:–154 °C
:0.653 g/cm³ (20 °C)
:203-523-4
:3,II
:M-Clarity™ quality level = MQ100
:–33 °C
:86.18 g/mol
:1208
:GHS02
:GHS08
:GHS07
:GHS09
:Ambient
:107-83-5
:C₆H₁₄
:MFCD00009406