



2-Methylpentane for synthesis, Sigma-Aldrich®
: Merck
:107-83-5
:
:C₆H₁₄
:MFCD00009406
:62 °C (1013 hPa)
:–154 °C
:0.653 g/cm³ (20 °C)
:–33 °C
:Ambient
:203-523-4
:1208
:3,II
:M-Clarity™ quality level = MQ200
:86.18 g/mol
:GHS02
:GHS08
:GHS07
:GHS09
- Solubility:0.14 g/l (25 °C)
- Melting Point:-154 °C
- Molar Mass:86.18 g/mol
- Boiling Point:60 °C (1013 hPa)
- Vapor Pressure:227 hPa (20 °C)
- Flash Point:-40 °C
- Refractive Index:1.3715 (20 °C, 589 nm)
- Explosion Limit:1.2 - 7.4 %(V)
- Density:0.65 g/cm3 (20 °C)
- Ignition Point:300 °C
- Synonyms:iso-Hexane, 1-Dimethylbutane
- Hill Formula:C6H14
- Chemical Formula:CH3CH2CH2CH(CH3)2
Frequently Bought Together