2-Methylpentane ≥99%
About this item
Purity: 99+% CAS No.: 107-83-5 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -40°(-40°F) Refractive Index: 1.3710 Odour: Petroleum-like Melting Point: -154° Boiling Point: 61-62° Beilstein Reference: 1730735 UN No.: 1208 MDL No.: MFCD00009406
- CAS Number:107-83-5
- Formula:C₆H₁₄
- MDL Number:MFCD00009406
- Molecular Weight:86.18 g/mol
- Boiling Pt:61…62 °C
- Melting Pt:–154 °C
- Density:0.653
- Flash Pt:–40 °C (–40 °F)
- Storage Temperature:Ambient
- EINECS:203-523-4
- UN:1208
- ADR:3,II
2 Options Available Below
Product Details & Documents
Purity: 99+% CAS No.: 107-83-5 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -40°(-40°F) Refractive Index: 1.3710 Odour: Petroleum-like Melting Point: -154° Boiling Point: 61-62° Beilstein Reference: 1730735 UN No.: 1208 MDL No.: MFCD00009406
- CAS Number:107-83-5
- Formula:C₆H₁₄
- MDL Number:MFCD00009406
- Molecular Weight:86.18 g/mol
- Boiling Pt:61…62 °C
- Melting Pt:–154 °C
- Density:0.653
- Flash Pt:–40 °C (–40 °F)
- Storage Temperature:Ambient
- EINECS:203-523-4
- UN:1208
- ADR:3,II
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



