Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

2-Methylheptane 99%

Irritant
Health
Environment
Flammable
2-Methylheptane 99%
Certificates
2-Methylheptane 99%

3 Options Available Below

Product Details & Documents
Synonyms:  Isoheptane
  • CAS Number:592-27-8
  • Formula:C₈H₁₈
  • MDL Number:MFCD00009518
  • Molecular Weight:114.23 g/mol
  • Boiling Pt:116 °C (761 mmHg)
  • Melting Pt:–108.9 °C
  • Density:0.69 g/cm³
  • Flash Pt:4 °C
  • Storage Temperature:Ambient
  • EINECS:209-747-9
  • UN:1262
  • ADR:3,II

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3930 to 1.3970 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 456,00 kr
    456,00 kr
    1 * 1 g
       456,00 kr

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1.170,00 kr
    1.170,00 kr
    1 * 5 g
       1.170,00 kr

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 4.320,00 kr
    4.320,00 kr
    1 * 25 g
       4.320,00 kr