2-Methylbutane ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: iso-Pentane
CAS Number:78-78-4
:
Formula:CH₃CH₂CH(CH₃)₂
MDL Number:MFCD00009338
Boiling Pt:30 °C
Melting Pt:–160 °C
Density:0.62
Molecular Weight:72.15 g/mol
Flash Pt:–57 °C (–70 °F)
EINECS:201-142-8
UN:1265
ADR:3 I
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together