Order Entry
Denmark
ContactUsLinkComponent
pH/mV/°C meter, handheld, 826
pH/mV/°C meter, handheld, 826
Catalog # 662-0413
Supplier:  METROHM
undefined
pH/mV/°C meter, handheld, 826
Catalog # 662-0413
Supplier:  METROHM

Specifications

  • Temperature range:
    −5...+250 (NTC); −150...+250 (Pt1000)
  • Description:
    Handheld pH/mV/°C meter 826 without carrying case
  • Model:
    826
  • Temperature accuracy:
    ±0,6 (NTC); ±0,2 (Pt1000)
  • Temperature resolution:
    0,1 °C
  • Compliance:
    GLP / IP 67
  • Connections:
    Infrared PC and printer connection
  • Data logging capacity:
    200 measuring values
  • Power supply:
    4×1,5 V batteries
  • mV accuracy:
    ±0,02
  • mV range:
    ±1200
  • mV resolution:
    0,1
  • pH accuracy:
    ±0,003
  • pH calibration:
    1, 2 or 3-point
  • pH range:
    −8...+22
  • pH resolution:
    0,001
  • Pk:
    1

Specifications

About this item

Handy and easy-to-use pH meter with rugged plastic housing for daily routine work in the laboratory.

  • Wireless infrared interface for printer and PC
  • User and sample identification
  • Automatic temperature compensation automatic buffer recognition
  • Three-point calibration with monitoring functions
  • Memory possibility for 200 measuring values with date, time and sample identification
Ordering information: 826 supplied as instrument alone (incl. carrying strap, 4×1,5 V batteries, hexagon key and user manua) or with a carrying case incl. Primatrode pH electrode, PE sleeve, sample/wash bottles, buffer solutions pH 4, 7 and 9, carrying strap and hexagon key.