

Nitro-tert-butane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:594-70-7
:
Formula:C₄H₉NO₂
MDL Number:MFCD00007393
:103.12 g/mol
Boiling Pt:126…127 °C
Melting Pt:27…31 °C
Density:0.950
Flash Pt:19 °C (66 °F)
Storage Temperature:Ambient
EINECS:209-851-4
UN:1325
ADR:4.1,II
:GHS02
:GHS07
Related Information
Frequently Bought Together