2-Chloro-2-methylbutane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:594-36-5
:
Formula:C₅H₁₁Cl
MDL Number:MFCD00000818
Boiling Pt:85…86 °C
Melting Pt:–73 °C
Density:0.866
Molecular Weight:106.6 g/mol
Flash Pt:–10 °C (14 °F)
Storage Temperature:Ambient
EINECS:209-836-2
UN:1107
ADR:3 II
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together