
1,5-Dibromo-3-methylpentane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4457-72-1
:
Formula:C₆H₁₂Br₂
MDL Number:MFCD00039200
:243.97 g/mol
Boiling Pt:233 °C
Density:1.58
Flash Pt:101 °C (214 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together