Order Entry
Denmark
ContactUsLinkComponent
 
 
 
 
 
n-Pentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
 
undefined
 
 
 
 
n-Pentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

 

  • Assay (GC):
    Min. 99 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Explosion Limit:
    ~8.3 %
  • Autoignition Temperature:
    500 °F

 

 

Frequently Bought Together