n-Pentane ≥98%
Purity: 98% CAS No.: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.626 Refractive Index: 1.3580 Appearance: Colourless Odour: Nearly Odourless Melting Point: -130° Boiling Point: 36° Merck Reference: 14,7116 UN No.: 1265 MDL No.: MFCD00009498
- :109-66-0
- :H₃C(CH₂)₃CH₃
- :MFCD00009498
- :72.15 g/mol
- :14,7116
- :36 °C
- :–130 °C
- :0.626
- :–49 °C (–57 °F)
- :Ambient
- :203-692-4
- :1265
- :3,II
- :01-2119459286-30
Purity: 98% CAS No.: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.626 Refractive Index: 1.3580 Appearance: Colourless Odour: Nearly Odourless Melting Point: -130° Boiling Point: 36° Merck Reference: 14,7116 UN No.: 1265 MDL No.: MFCD00009498
- :109-66-0
- :H₃C(CH₂)₃CH₃
- :MFCD00009498
- :72.15 g/mol
- :14,7116
- :36 °C
- :–130 °C
- :0.626
- :–49 °C (–57 °F)
- :Ambient
- :203-692-4
- :1265
- :3,II
- :01-2119459286-30