Spillage trays
About this item
Made of HDPE, white or black (electrically conductive). Containers for all purposes.
- Electrically conductive available
- Available with base insert; the canisters always stay dry
- Anti-static mat and grund cable optionally available
Suitable for a wide range of canisters. Collect liquid in case of a leakage. The SCAT collection trays prevent the uncontrolled spread of hazardous liquids in the event of damage to a disposal container. The HDPE-EC conductive tray also prevents the build-up of static charge, which prevents the ignition of explosive substances.
5 Options Available Below
Product Details & Documents
Made of HDPE, white or black (electrically conductive). Containers for all purposes.
- Electrically conductive available
- Available with base insert; the canisters always stay dry
- Anti-static mat and grund cable optionally available
Suitable for a wide range of canisters. Collect liquid in case of a leakage. The SCAT collection trays prevent the uncontrolled spread of hazardous liquids in the event of damage to a disposal container. The HDPE-EC conductive tray also prevents the build-up of static charge, which prevents the ignition of explosive substances.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Product Family Options
Product Information
- ColourTypeW×D×H ext.W×D×H int.AvailabilityPrice
- 590-0809Collecting tray, PEDH materialCollecting tray, PEDH materialWhitenot conductive, without base insert300×400×170 mm235×335×160 mmLog in for availability1 * 1 Items620,00 kr
- SCTV117984Collecting tray, PEDH materialCollecting tray, PEDH materialWhiteNot conductive, without base insert340×465×210 mm290×385×200 mmLog in for availability1 * 1 Items620,00 kr
- 216-0924Collecting tray, PEDH materialCollecting tray, PEDH materialBlackElectrically conductive, without base insert325×225×215 mm300×200×200 mmLog in for availability1 * 1 Items1.770,00 kr
- 216-0925Collecting tray with base insert, PEDH-EC materialCollecting tray with base insert, PEDH-EC materialBlackElectrically conductive, with base insert355×445×135 mm285×385×95 mmLog in for availability1 * 1 Items2.390,00 kr
- 216-0926Collecting tray with base insert, PEDH-EC materialCollecting tray with base insert, PEDH-EC materialBlackElectrically conductive, with base insert365×490×240 mm295×415×200 mmLog in for availability1 * 1 Items2.650,00 kr
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



