Organic reference standard, Tris(2-chloroethyl) phosphate 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL17894300CY
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:115-96-8
- Description:Tris(2-chloroethyl) phosphate 10 µg/ml in cyclohexane
- Formula:C₆H₁₂Cl₃O₄P
- Description:Tris(2-chloroethyl) phosphate