Order Entry
Denmark
Orders LinkContactUsLinkComponent
n-Hexane, Supelco®

n-Hexane, Supelco®

Glass bottle, 10 mL
Catalog # SUPL52750-10ML
Flammable
Health
Irritant
Environment
Supplier:   Merck
n-Hexane, Supelco®
Catalog # SUPL52750-10ML
Supplier:  Merck
CAS Number:  110-54-3
n-Hexane, Supelco®

Glass bottle, 10 mL

CAS Number:  110-54-3
Price
1.180,00 kr
Log in for availability
1 * 10 mL
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94.3 °C
  • Density:0.659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.