n-Decane ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-18-5
:
Formula:H₃C(CH₂)₈CH₃
MDL Number:MFCD00008954
:142.28 g/mol
Boiling Pt:172…174 °C
Melting Pt:–30 °C
Density:0.730
Flash Pt:46 °C (115 °F)
Storage Temperature:Ambient
EINECS:204-686-4
UN:2247
ADR:3,III
:GHS08
:GHS02
Related Information
Frequently Bought Together