n-Decane 99+%, pure
:124-18-5
:
:H₃C(CH₂)₈CH₃
:MFCD00008954
:142.28 g/mol
:174 °C (760 mmHg)
:–30 °C
:0.735 g/cm³
:46 °C
:Ambient
:204-686-4
:2247
:3,III
:GHS08
:GHS02
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4100 to 1.4130 (20°C, 589 nm)
- GC:≥99.0 %
- Composition of fatty acids:5.0 to 14.0 % (GC) Palmitic acid
- Acid value:≤0.5 mg KOH/g
- Peroxide value:≤5 meq O2/kg
- Unsaponifiable matter:≤1 %
Frequently Bought Together