

Methane
Catalog # REST34454
Supplier: Restek
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:16.04 g/mol
Merck Index:12,06019
Boiling Pt:–161.5 °C (1013 hPa)
Melting Pt:–182.5 °C
Density:0.717 g/cm³ (20 °C)
Flash Pt:–221 °C
Storage Temperature:Ambient
EINECS:200-812-7
UN:1971
ADR:2,
:GHS02
:GHS04
CAS Number:74-82-8
Formula:CH₄