1,6-Hexanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:629-11-8
:
Formula:C₆H₁₄O₂
MDL Number:MFCD00002985
:118.18 g/mol
Merck Index:13,04709
Boiling Pt:250 °C (1013 hPa)
Melting Pt:39…42 °C
Density:1.12 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:211-074-0
Specification Test Results
- Solubility:5000 g/l (20 °C)
- Melting Point:39 - 42 °C
- Molar Mass:118.17 g/mol
- Bulk Density:530 kg/m3
- Boiling Point:253 - 260 °C
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:147 °C
- Explosion Limit:6.6 - 16 %(V)
- Density:1.12 g/cm3 (20 °C)
- pH:7.6 (900 g/l, H2O, 20 °C)
- Ignition Point:320 °C
Specification Test Results
Frequently Bought Together