Order Entry
Denmark
Orders LinkContactUsLinkComponent
Methanol

Methanol

1
Catalog # EDQMY0000428
Flammable
Toxic
Health
Supplier:   EDQM
Methanol
Catalog # EDQMY0000428
Supplier:  EDQM
CAS Number:  67-56-1
Methanol
CAS Number:  67-56-1
Price
1.920,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications

Specifications

  • Pk:
    1

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.