1-Pentanol, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:71-41-0
:
Formula:H₃C(CH₂)₃CH₂OH
MDL Number:MFCD00002977
:88.15 g/mol
Merck Index:13,07195
Boiling Pt:137.9 °C (1013 hPa)
Melting Pt:–79 °C
Density:0.81 g/cm³ (20 °C)
EINECS:200-752-1
UN:1105
ADR:3,III
:M-Clarity™ quality level = MQ100
Flash Pt:48 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together