n-Amylalkohol
Catalog # PROOCHE138
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pakning:Glass bottle
- Pakn:2 mL
- Seq:2145
Specifications
About this item
:
:88.15 g/mol
Merck Index:13,07195
Boiling Pt:137,9 °C (1013 hPa)
Melting Pt:–79 °C
Density:0,81 g/cm³ (20 °C)
Flash Pt:48 °C
Storage Temperature:Stue temperatur
EINECS:200-752-1
UN:1105
ADR:3,III
:GHS02
:GHS07
CAS Number:71-41-0
Formula:H₃C(CH₂)₃CH₂OH
MDL Number:MFCD00002977