1-Octadecanol for synthesis
Supplier: Merck
Certificates
About this item
Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
CAS Number:112-92-5
:
Formula:H₃C(CH₂)₁₆CH₂OH
MDL Number:MFCD00002823
:270.5 g/mol
Merck Index:12,08960
Boiling Pt:336 °C (1013 hPa)
Melting Pt:56…57 °C
Density:0.805…0.815 g/cm³ (60 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-017-6
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Hill Formula:C₁₈H₃₈O
- Molar Mass:270.49 g/mol
- Boiling point:330 - 360 °C (1013 hPa)
- Density:0.805 - 0.815 g/cm³ (60 °C)
- Flash point:195 °C
- Ignition temperature:230 °C DIN 51794
- Melting Point:55 - 60 °C
- Vapor pressure:- 1 hPa (20 °C)
- Bulk density:300 kg/m³
- Solubility:(20 °C) practically insoluble
- LD 50 oral:LD50 Rat 2000 mg/kg
- Storage class:10 - 13 Other liquids and solids
- WGK:NWG not water endangering
Specification Test Results
Related Information
Frequently Bought Together