1-Hexadecanol
Om denne vare
Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.
Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.
Easily soluble in diethyl ether, soluble in acetone, chloroform; slightly soluble in alcohol.
Store at +4 °C
- CAS nummer:36653-82-4
- Formel:H₃C(CH₂)₁₄CH₂OH
- MDL nummer:MFCD00004760
- Molecular Weight:242.45 g/mol
- Merck Index:13,02037
- Kogende Pt:305…330 °C (1013 hPa)
- Smeltende Pt:46…52 °C
- Tæthed:0.81 g/cm³ (20 °C)
- Flash Pt:>110 °C
- Opbevaringstemperatur:Ambient
- EINECS:253-149-0
2 tilgængelige muligheder nedenfor
Product Details & Documents
Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.
Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.
Easily soluble in diethyl ether, soluble in acetone, chloroform; slightly soluble in alcohol.
Store at +4 °C
- CAS nummer:36653-82-4
- Formel:H₃C(CH₂)₁₄CH₂OH
- MDL nummer:MFCD00004760
- Molecular Weight:242.45 g/mol
- Merck Index:13,02037
- Kogende Pt:305…330 °C (1013 hPa)
- Smeltende Pt:46…52 °C
- Tæthed:0.81 g/cm³ (20 °C)
- Flash Pt:>110 °C
- Opbevaringstemperatur:Ambient
- EINECS:253-149-0
Product Family Options
Produktinformation
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



