1-Hexadecanol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 36653-82-4 Molecular Formula: CH3(CH2)15OH Formula Weight: 242.45 Flash Point: 135°(275°F) Physical Form: Crystalline Density: 0.817 Melting Point: 47-51° Boiling Point: 190°/15mm Solubility: Soluble in alcohol, chloroform, ether Beilstein Reference: 1748475 Merck Reference: 14,2028 MDL No.: MFCD00004760
Synonyms: Cetyl alcohol, Palmityl alcohol
- CAS Number:36653-82-4
- Formula:H₃C(CH₂)₁₄CH₂OH
- MDL Number:MFCD00004760
- Molecular Weight:242.45 g/mol
- Merck Index:14,2028
- Boiling Pt:190 °C (15 mmHg)
- Melting Pt:47…51 °C
- Density:0.817
- Flash Pt:135 °C (275 °F)
- Storage Temperature:Ambient
- EINECS:253-149-0
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 36653-82-4 Molecular Formula: CH3(CH2)15OH Formula Weight: 242.45 Flash Point: 135°(275°F) Physical Form: Crystalline Density: 0.817 Melting Point: 47-51° Boiling Point: 190°/15mm Solubility: Soluble in alcohol, chloroform, ether Beilstein Reference: 1748475 Merck Reference: 14,2028 MDL No.: MFCD00004760
Synonyms: Cetyl alcohol, Palmityl alcohol
- CAS Number:36653-82-4
- Formula:H₃C(CH₂)₁₄CH₂OH
- MDL Number:MFCD00004760
- Molecular Weight:242.45 g/mol
- Merck Index:14,2028
- Boiling Pt:190 °C (15 mmHg)
- Melting Pt:47…51 °C
- Density:0.817
- Flash Pt:135 °C (275 °F)
- Storage Temperature:Ambient
- EINECS:253-149-0



