1-Hexadecanol 96%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Cetyl alcohol, Palmityl alcohol
- CAS Number:36653-82-4
- Formula:H₃C(CH₂)₁₄CH₂OH
- MDL Number:MFCD00004760
- Molecular Weight:242.45 g/mol
- Merck Index:13,02037
- Boiling Pt:344 °C (760 mmHg)
- Melting Pt:47…50 °C
- Density:0,81 g/cm³ (20 °C)
- Flash Pt:>157 °C
- Storage Temperature:Ambient
- EINECS:253-149-0
3 Options Available Below
Product Details & Documents
Synonyms: Cetyl alcohol, Palmityl alcohol
- CAS Number:36653-82-4
- Formula:H₃C(CH₂)₁₄CH₂OH
- MDL Number:MFCD00004760
- Molecular Weight:242.45 g/mol
- Merck Index:13,02037
- Boiling Pt:344 °C (760 mmHg)
- Melting Pt:47…50 °C
- Density:0,81 g/cm³ (20 °C)
- Flash Pt:>157 °C
- Storage Temperature:Ambient
- EINECS:253-149-0
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Powder or flakes or pellets
- Infrared spectrum:Conforms
- GC:≥95 %



