Order Entry
Denmark
ContactUsLinkComponent
 
 
 
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 
undefined
 
 
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 
 
 

 

  • Assay:
    Min. 99.7 %
  • Assay (Ionol/BHT):
    4.0 - 10.0 ppm
  • Aldehydes:
    Not detected by GC
  • Insolubility in water:
    Passes test
  • IR Spectrum:
    Passes test
  • Acidity:
    Max. 0.0001 meq/g
  • Boiling point:
    34 - 35 °C
  • Colouration:
    Max. 10 APHA
  • Relative density:
    0.714 - 0.715
  • Substances coloured by H2SO4:
    Max. 10 APHA
  • Acetone:
    Max. 10 ppm
  • Aldehydes + ketones (as HCHO):
    Max. 5 ppm
  • Carbonyl compounds (as HCHO):
    Max. 10 ppm
  • Ethanol:
    Max. 0.05 %
  • Evaporation residue:
    Max. 5 ppm
  • Methanol:
    Max. 0.02 %
  • Peroxides (as H2O2):
    Max. 0.15 ppm
  • Water:
    Max. 300 ppm
  • Al (Aluminium):
    Max. 0.5 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.05 ppm
  • Co (Cobalt):
    Max. 0.02 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • K (Potassium):
    Max. 0.05 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.02 ppm
  • Na (Sodium):
    Max. 0.1 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Sr (Strontium):
    Max. 0.02 ppm
  • Zn (Zinc):
    Max. 0.1 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

 

Frequently Bought Together