Order Entry
Denmark
ContactUsLinkComponent
1-Nitropropane for synthesis, Sigma-Aldrich®
Flammable
Irritant
1-Nitropropane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
1-Nitropropane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    14 g/l (20 °C)
  • Melting Point:
    -108 °C
  • Molar Mass:
    89.09 g/mol
  • Boiling Point:
    131 - 132 °C (1013 hPa)
  • Vapor Pressure:
    7.6 hPa (20 °C)
  • Flash Point:
    36 °C
  • Refractive Index:
    1.4016 (20 °C, 589 nm)
  • Explosion Limit:
    2.2 - 11.0 %(V)
  • Density:
    0.99 g/cm3 (20 °C)
  • pH:
    6.0 (0.9 g/l, H2O, 20 °C)
  • Ignition Point:
    420 °C

Specification Test Results

Related Information

Frequently Bought Together