

1-Nitropropane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-03-2
:
Formula:C₃H₇NO₂
MDL Number:MFCD00007407
:89.09 g/mol
Merck Index:14,6627
Boiling Pt:131…132 °C
Melting Pt:–108 °C
Density:0.998
Flash Pt:34 °C (93 °F)
Storage Temperature:Ambient
EINECS:203-544-9
UN:2608
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together