


Nonylamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-20-9
:
Formula:H₃C(CH₂)₇CH₂NH₂
MDL Number:MFCD00008249
:143.27 g/mol
Boiling Pt:202 °C (760 mmHg)
Melting Pt:–1 °C
Density:0.784 g/cm³
Flash Pt:63 °C
Storage Temperature:Ambient
EINECS:203-945-9
UN:2735
ADR:8,II
:GHS07
:GHS05
:GHS09
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4320 to 1.4350 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together