Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Dichloromethane, Supelco®

Health
Irritant
Supplier:   Merck
Dichloromethane, Supelco®
Supplier:  Merck
Certificates
Dichloromethane, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39.8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1.322 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-838-9
  • UN:1593
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 519,00 kr
    519,00 kr
    1 * 100 mL
       519,00 kr

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1.080,00 kr
    1.080,00 kr
    1 * 500 mL
       1.080,00 kr