Order Entry
Denmark
ContactUsLinkComponent
Ammonium acetate ≥97%, GPR RECTAPUR®
Ammonium acetate ≥97%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Ammonium acetate ≥97%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 97 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.05 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together