Order Entry
Denmark
Orders LinkContactUsLinkComponent
Potassium acetate

Potassium acetate

Marketsource
Supplier:   Roth Carl
Potassium acetate
Supplier:  Roth Carl
Marketsource
Certificates
Potassium acetate
Potassium acetate

3 Options Available Below

Product Details & Documents
Synonyms:  Acetic acid potassium salt
  • CAS Number:127-08-2
  • Formula:CH₃COOK
  • MDL Number:MFCD00012458
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,07687
  • Melting Pt:304 °C
  • Density:1.57 g/cm³ (25 °C)
  • Flash Pt:>250 °C
  • Storage Temperature:Ambient
  • EINECS:204-822-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 317,37 kr
    317,37 kr
    1 * 500 g
       317,37 kr

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 581,73 kr
    581,73 kr
    1 * 1 kg
       581,73 kr

    Specifications:
    Marketsource
    Marketsource
  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 9.334,54 kr
    9.334,54 kr
    1 * 25 kg
       9.334,54 kr

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.