Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Myristic acid

Glass bottle for solids, 1 kg
Katalog # ROTH6469.1
Irriterende
Marketsource
Leverandør:   Roth Carl
Myristic acid
Katalog # ROTH6469.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  544-63-8
Myristic acid

Glass bottle for solids, 1 kg

CAS nummer:  544-63-8
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Tetradecylic acid, Tetradecanoic acid
  • CAS nummer:544-63-8
  • Formel:C₁₄H₂₈O₂
  • MDL nummer:MFCD00002744
  • Molecular Weight:228.38 g/mol
  • Merck Index:12,06416
  • Kogende Pt:319 °C (1013 hPa)
  • Smeltende Pt:51…54 °C
  • Tæthed:0.8622 g/cm³ (20 °C)
  • Flash Pt:>165 °C
  • Opbevaringstemperatur:Freezer
  • EINECS:208-875-2
Specifikationer

Specifikationer

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifikationer