Myristic acid
Catalog #
Product Details & Documents
Synonyms: Tetradecylic acid, Tetradecanoic acid
- CAS Number:544-63-8
- Formula:C₁₄H₂₈O₂
- MDL Number:MFCD00002744
- Molecular Weight:228.38 g/mol
- Merck Index:12,06416
- Boiling Pt:319 °C (1013 hPa)
- Melting Pt:51…54 °C
- Density:0.8622 g/cm³ (20 °C)
- Flash Pt:>165 °C
- Storage Temperature:Freezer
- EINECS:208-875-2



