Myristic acid ≥98%
Leverandør: Thermo Fisher Scientific
Gå til:
Om denne vare
Purity: 98% CAS No.: 544-63-8 Molecular Formula: CH3(CH2)12CO2H Formula Weight: 228.38 Flash Point: 175°(347°F) Melting Point: 53-56° Boiling Point: 326° Beilstein Reference: 508624 Merck Reference: 14,6333 MDL No.: MFCD00002744
Synonymer: Tetradecylic acid, Tetradecanoic acid
- CAS nummer:544-63-8
- Formel:C₁₄H₂₈O₂
- MDL nummer:MFCD00002744
- Molecular Weight:228.38 g/mol
- Merck Index:14,6333
- Kogende Pt:326 °C
- Smeltende Pt:53…56 °C
- Tæthed:0.862
- Flash Pt:175 °C (347 °F)
- Opbevaringstemperatur:Ambient
- EINECS:208-875-2
3 tilgængelige muligheder nedenfor
Product Details & Documents
Purity: 98% CAS No.: 544-63-8 Molecular Formula: CH3(CH2)12CO2H Formula Weight: 228.38 Flash Point: 175°(347°F) Melting Point: 53-56° Boiling Point: 326° Beilstein Reference: 508624 Merck Reference: 14,6333 MDL No.: MFCD00002744
Synonymer: Tetradecylic acid, Tetradecanoic acid
- CAS nummer:544-63-8
- Formel:C₁₄H₂₈O₂
- MDL nummer:MFCD00002744
- Molecular Weight:228.38 g/mol
- Merck Index:14,6333
- Kogende Pt:326 °C
- Smeltende Pt:53…56 °C
- Tæthed:0.862
- Flash Pt:175 °C (347 °F)
- Opbevaringstemperatur:Ambient
- EINECS:208-875-2
Documents
Product Family Options
Produktinformation
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



