Propionic acid
Catalog # EHERC16493000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:74.08 g/mol
Merck Index:13,07917
Boiling Pt:141 °C (1013 hPa)
Melting Pt:–21 °C
Density:0.99 g/cm³ (20 °C)
Flash Pt:49 °C
Storage Temperature:Ambient
EINECS:201-176-3
UN:3463
ADR:8,II
REACH:01-2119486971-24
:GHS05
:GHS07
CAS Number:79-09-4
Formula:C₃H₆O₂
MDL Number:MFCD00002756