Order Entry
Denmark
Orders LinkContactUsLinkComponent
Valeric acid, Supelco®

Valeric acid, Supelco®

Corrosive
Supplier:   Merck
Valeric acid, Supelco®
Supplier:  Merck
Certificates
Valeric acid, Supelco®
Valeric acid, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  Pentanoic acid
  • CAS Number:109-52-4
  • Formula:C₅H₁₀O₂
  • MDL Number:MFCD00004413
  • Molecular Weight:102.13 g/mol
  • Merck Index:12,10042
  • Boiling Pt:186 °C (1013 hPa)
  • Melting Pt:–32 °C
  • Density:1.11 g/cm³ (20 °C)
  • Flash Pt:89 °C
  • Storage Temperature:Ambient
  • UN:3265
  • ADR:8,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-677-2

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.