Caprylic acid ≥99.5%, liquid
:124-07-2
:
:C₈H₁₆O₂
:MFCD00004429
:144.21 g/mol
:13,01771
:237 °C (lit.)
:15…17 °C (lit.)
:0.91 g/cm³ (20 °C) (lit.)
:130 °C
:Ambient
:204-677-5
:3265
:8,III
:GHS06
:GHS05
- Identity Test :Passes
- Purity:99.6%
- FTIR:Conforms to Standard
Frequently Bought Together