Stearic acid, powder, TECHNICAL
Supplier: VWR Chemicals
Om denne vare
Synonymer: Octadecanoic acid, Octadecylic acid
- CAS nummer:57-11-4
- Formel:C₁₈H₃₆O₂
- MDL nummer:MFCD00002752
- Molecular Weight:284.48 g/mol
- Merck Index:13,08882
- Kogende Pt:361 °C (1013 hPa)
- Smeltende Pt:67 °C
- Tæthed:0.94 g/cm³ (20 °C)
- Flash Pt:196 °C (closed cup)
- Opbevaringstemperatur:Refrigerator
- EINECS:200-313-4
- NÅ:01-2119543894-28
2 tilgængelige muligheder nedenfor
Product Details & Documents
Synonymer: Octadecanoic acid, Octadecylic acid
- CAS nummer:57-11-4
- Formel:C₁₈H₃₆O₂
- MDL nummer:MFCD00002752
- Molecular Weight:284.48 g/mol
- Merck Index:13,08882
- Kogende Pt:361 °C (1013 hPa)
- Smeltende Pt:67 °C
- Tæthed:0.94 g/cm³ (20 °C)
- Flash Pt:196 °C (closed cup)
- Opbevaringstemperatur:Refrigerator
- EINECS:200-313-4
- NÅ:01-2119543894-28
Certifikater
Søg efter partinummerSpecifications for Product Family
Specifikationstestresultater
- Stearic and palmitic acid:Min. 90 %
Product Family Options
Produktinformation
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



